Orochem Orosil Gold C18-HC and C18-Polar HPLC Columns
Ideal for separation of polar, semi-polar and nonpolar compounds at low to medium pH
  • High carbon-loaded phase and fully endcapped
  • Constructed with high purity silica gel
These Orochem Orosil Gold C18 HPLC columns are uniquely designed for the separation of polar, semi-polar and nonpolar compounds at low to medium pH. Stationary phases use high purity silica gel.
Orosil Gold C18-HC stationary phase columns are high carbon-loaded phase containing 20% carbon and are fully endcapped. These columns feature a pH range of 2 to 8 pH.
Orosil Gold C18-Polar stationary phase columns are high carbon-loaded phase containing 19% carbon and feature a proprietary polar endcap. Columns are designed for difficult separations of polar and semi-polar compounds and have superior selectivity of basic compounds. The combination of high carbon-loading and polar endcapping produces a phase that contains high hydrophobicity and polarity making columns compatible with 100% aqueous phases. These columns feature a pH range of 2 to 9 pH.
$607.50 - $761.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Length (mm) Column Particle Size (µm) Phase
Availability Pricing
Orochem OroSil™ Gold HPLC Column, C18-HC, 150 mm L x 4.6 mm ID EW-06486-27 Mfr # O3FI-027
COMPARE
150 3 C18-HC
$616.00USD / Each
Orochem OroSil™ Gold HPLC Column, C18-HC, 150 mm L x 4.6 mm ID EW-06486-28 Mfr # O5FI-027
COMPARE
150 5 C18-HC
$607.50USD / Each
Orochem OroSil™ Gold HPLC Column, C18-HC, 250 mm L x 4.6 mm ID EW-06486-29 Mfr # O3HI-027
COMPARE
250 3 C18-HC
$761.00USD / Each
Orochem OroSil™ Gold HPLC Column, C18-HC, 250 mm L x 4.6 mm ID EW-06486-30 Mfr # O5HI-027
COMPARE
250 5 C18-HC
$723.00USD / Each
Orochem OroSil™ Gold HPLC Column, C18-Polar, 150 mm L x 4.6 mm ID EW-06486-31 Mfr # O3FI-024
COMPARE
150 3 C18-Polar
$616.00USD / Each
Orochem OroSil™ Gold HPLC Column, C18-Polar, 150 mm L x 4.6 mm ID EW-06486-32 Mfr # O5FI-943
COMPARE
150 5 C18-Polar
$607.50USD / Each
Orochem OroSil™ Gold HPLC Column, C18-Polar, 250 mm L x 4.6 mm ID EW-06486-33 Mfr # O5HI-943
COMPARE
250 5 C18-Polar
$751.00USD / Each
Showing 1- 7 of 7