Orochem Reliasil™ Silica HPLC Columns
Rugged design with enhanced chemical stability and efficiency
  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for Silica phase
The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume.Standard columns feature a pore size of 90 Å.
$598.50 - $5,772.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Orochem Reliasil™ HPLC Column, Silica, 3 µm, 100 mm L x 3.0 mm ID, 90Å EW-06486-70 Mfr # R3DG-100
COMPARE
3 100 3
$645.00USD / Each
Orochem Reliasil™ HPLC Column, Silica, 5 µm, 100 mm L x 3.0 mm ID, 90Å EW-06486-71 Mfr # R5DG-120
COMPARE
3 100 5
$598.50USD / Each
Orochem Reliasil™ HPLC Column, Silica, 3 µm, 150 mm L x 4.6 mm ID, 90Å EW-06486-72 Mfr # R3FI-100
COMPARE
4.6 150 3
$618.00USD / Each
Orochem Reliasil™ HPLC Column, Silica, 5 µm, 150 mm L x 4.6 mm ID, 90Å EW-06486-73 Mfr # R5FI-120
COMPARE
4.6 150 5
$618.00USD / Each
Orochem Reliasil™ HPLC Column, Silica, 5 µm, 250 mm L x 4.6 mm ID, 90Å EW-06486-74 Mfr # R5HI-120
COMPARE
10 250 5
$706.50USD / Each
Orochem Reliasil™ HPLC Column, Silica, 10 µm, 250 mm L x 10 mm ID, 90Å EW-06486-75 Mfr # R10HL-140
COMPARE
10 250 10
$3,176.00USD / Each
Orochem Reliasil™ HPLC Column, Silica, 5 µm, 250 mm L x 10 mm ID, 90Å EW-06486-76 Mfr # R9HL-140
COMPARE
10 250 5
$1,357.00USD / Each
Orochem Reliasil™ HPLC Column, Silica, 10 µm, 250 mm L x 21 mm ID, 90Å EW-06486-77 Mfr # R10HT-140
COMPARE
21 250 10
$5,772.00USD / Each
Orochem Reliasil™ HPLC Column, Silica, 5 µm, 250 mm L x 21 mm ID, 90Å EW-06486-78 Mfr # R9HM-140
COMPARE
21 250 5
$3,395.00USD / Each
Showing 1- 9 of 9