Ovation Macrovolume Electronic Pipettors
Proven ergonomically correct pipettor is comfortable and easy-to-use!
  • Minimize contact pressure, tension and exertion
  • Eliminate the need for organizers, the Ovation pipettor can stand on its own!
  • Flawless tipping and detipping, and simple menu make it easy to use
The Ovation design changes the concept of liquid transfer by reducing the causes of pain, fatigue, and stress from pipetting. Work for hours without tension in the shoulder or elbow. Never over-rotate your wrist or hand—the Ovation pipettor was formulated to keep your hand in the neutral position recommended by ergonomic experts. This macro pipettor eliminates the uncomfortable and tiring postures that come with large-volume liquid handling, and can easily fit under a working hood.
Fully electronic pipettor provides a broad menu of liquid handling functions and maximizes pipetting efficiency. Return to the factory calibration setting with just the push of a keypad button, making in-lab calibration simple. Model sold below is designed for right-handed users; contact our Application Specialists for left-handed model.
The disposable, polypropylene tips offer easy access into cell culture flasks, tubes, and bottles. The pipettor also accepts plastic and glass pipettes when extra-long length is needed to reach the bottom of tall vessels.
$884.00 - $895.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Min Volume (mL) Description
Availability Pricing
VistaLab Macro10 Macro Adjustable Volume Pipette, 200 uL to 10 mL EW-06197-42 Mfr # 1065-10ML
COMPARE
10 0.2
$884.00USD / Each
VistaLab Macro5 Macro Adjustable Volume Pipette, 100 uL to 5 mL EW-06199-67 Mfr # 1065-05ML
COMPARE
5 0.1
$895.00USD / Each