Oxygen (O2) Gas Mass Flowmeters with Metal bodies
Precisely measure flow rates and totals
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced straight-tube sensor that ensures accurate and repeatable
results. Gas flow measurements are unaffected by moderate temperature and pressure variations
at the inlet. The meter also allows a four-point calibration across the flow range to improve meter
linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 20 mA signal; an analogto-
RS converter is available for data collection and analysis on your PC.
All meters include a detachable 3 1/2-digit LCD that can be tilted up to 90° for easy reading.
Remotely mount the display up to three feet using extension cable 32662-70. The optional totalizer
32650-22 displays accumulated flow. The meter is protected from polarity reversal or short circuit
by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum,
brass, and Viton®; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77°F (15 to 25°C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77°F (15 to 25°C), add ±0.15%/°C full-scale.
$650.00 - $1,656.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Process Connection Wetted Materials
Availability Pricing
Aalborg GFMS-010034 Thermal Gas Mass Flowmeter, to 50 sccm O2, Aluminum body w/ display
Buy More Save More
EW-32649-04 Mfr # GFMS-010034
COMPARE
0.05 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg GFMS-010035 Thermal Gas Mass Flowmeter, to 100 sccm O2, Aluminum body w/ display
Buy More Save More
EW-32649-06 Mfr # GFMS-010035
COMPARE
0.1 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg GFMS-010336 Thermal Gas Mass Flowmeter, to 500 sccm O2, Aluminum body w/ display
Buy More Save More
EW-32649-10 Mfr # GFMS-010336
COMPARE
0.5 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg GFMS-010038 Thermal Gas Mass Flowmeter, to 1 L/min O2, Aluminum body w/ display
Buy More Save More
EW-32649-12 Mfr # GFMS-010038
COMPARE
1 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg GFMS-010039 Thermal Gas Mass Flowmeter, to 2 L/min O2, Aluminum body w/ display
Buy More Save More
EW-32649-14 Mfr # GFMS-010039
COMPARE
2 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg GFMS-010040 Thermal Gas Mass Flowmeter, to 5 L/min O2, Aluminum body w/ display
Buy More Save More
EW-32649-16 Mfr # GFMS-010040
COMPARE
5 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 5 L/min O2, 316SS Body
Buy More Save More
EW-32649-66 Mfr # GFMS-010055
COMPARE
5 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 15 L/min O2, 316SS Body
Buy More Save More
EW-32649-68 Mfr # GFMS-011286
COMPARE
15 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,222.00USD / Each
Aalborg GFMS-010554 Thermal Gas Mass Flowmeter, to 200 L/min O2, 316SS body w/ display
Buy More Save More
EW-32659-02 Mfr # GFMS-010554
COMPARE
200 3/8" Compression Fittings 316 Stainless Steel, Viton
$1,656.00USD / Each
Showing 1- 9 of 9