This product is now discontinued, View Related Products

Pacific Scientific OECO Gauss - Tesla Meter Accessories

Probes compatible with your Pacific Scientific meter.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Resistance Accuracy Resistance Range Resistance Resolution
Availability Pricing
Pacific Scientific-OECO SAH17-1904 Axial Probe, 4" for model 5170 gauss meter EW-20001-30 Mfr # SAH17-1904
COMPARE
Call For Price
Pacific Scientific-OECO STD18-0404 Transverse Probe, 4" for model 5180 Gauss Meter EW-20001-31 Mfr # STD18-0404
COMPARE
Call For Price
Pacific Scientific-OECO STH17-0404 (121704) Transverse Probe, model , 4" L EW-20043-27 Mfr # STH17-0404 (121704)
COMPARE
Call For Price
Pacific Scientific-OECO SAD18-1904 (121802) Axial Probe, model SAD18-1904, 4" L EW-20043-28 Mfr # SAD18-1904 (121802)
COMPARE
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat