Panel-Mount Compression Inserts 57171
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
$16.30 - $252.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
CPC (Colder) PMCD4004 Quick-Disconnect Panel-Mount Compression Insert; Valved, 1/4" Tube OD, 1/8" Flow Size
Buy More Save More
EW-06361-32 Mfr # PMCD4004
COMPARE
Acetal
$16.30USD / Each
CPC (Colder) PMC4004 Panel-Mount Compression Insert; Straight-through, 1/4" Tube OD, 1/8" Flow Size; 25/PK EW-31307-08 Mfr # PMC4004
COMPARE
Acetal
$252.00USD / Pkg of 25