Panel-Mount Hose Barb Bodies with 1/4" Flow
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
$16.70 - $377.00USD / Pkg of 25
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) PLCD16004 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/4" ID, 1/1" Flow, Acetal
Buy More Save More
EW-06360-55 Mfr # PLCD16004
COMPARE
1/4 Acetal Quick-Disconnect x Barb, Valved
$19.30USD / Each
CPC (Colder) PLCD16005 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Valved, 5/16" Tube ID, Acetal
Buy More Save More
EW-06360-57 Mfr # PLCD16005
COMPARE
5/16 Acetal Quick-Disconnect x Barb, Valved
$19.60USD / Each
CPC (Colder) PLCD16006 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Valved, 3/8" Tube ID, Acetal
Buy More Save More
EW-06360-60 Mfr # PLCD16006
COMPARE
3/8 Acetal Quick-Disconnect x Barb, Valved
$19.30USD / Each
CPC (Colder) PLC16004 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 1/4" Tube ID, 1/4" Flow Size
Buy More Save More
EW-06361-46 Mfr # PLC16004
COMPARE
1/4 Acetal Quick-Disconnect x Barb, Nonvalved
$17.60USD / Each
CPC (Colder) PLC16006 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 3/8" Tube ID, 1/4" Flow Size
Buy More Save More
EW-06361-48 Mfr # PLC16006
COMPARE
3/8 Acetal Quick-Disconnect x Barb, Nonvalved
$16.70USD / Each
CPC (Colder) PLCD1600412 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/4" ID, 1/4" Flow, PP
Buy More Save More
EW-06364-55 Mfr # PLCD1600412
COMPARE
1/4 Polypropylene Quick-Disconnect x Barb, Valved
$20.25USD / Each
CPC (Colder) PLCD1600612 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 3/8" Tube ID, PP
Buy More Save More
EW-06364-60 Mfr # PLCD1600612
COMPARE
3/8 Polypropylene Quick-Disconnect x Barb, Nonvalved
$21.50USD / Each
CPC (Colder) PLC16005 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 5/16" Tube ID, 1/4" Flow Size; 25/PK EW-31307-38 Mfr # PLC16005
COMPARE
5/16 Acetal Quick-Disconnect x Barb, Nonvalved
$377.00USD / Pkg of 25
Showing 1- 8 of 8