Panel-Mount Hose Barb Bodies with 1/8" Flow
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
$13.60 - $301.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) PMCD1601 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Valved, 1/16" Tube ID, Acetal
Buy More Save More
EW-06360-09 Mfr # PMCD1601
COMPARE
1/16 Acetal Quick-Disconnect x Barb, Valved
$15.70USD / Each
CPC (Colder) PMCD1602 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Valved, 1/8" Tube ID, Acetal
Buy More Save More
EW-06360-10 Mfr # PMCD1602
COMPARE
1/8 Acetal Quick-Disconnect x Barb, Valved
$16.90USD / Each
CPC (Colder) PMCD1603 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; Valved, 3/16" Tube ID, Acetal
Buy More Save More
EW-06360-12 Mfr # PMCD1603
COMPARE
3/16 Acetal Quick-Disconnect x Barb, Valved
$15.70USD / Each
CPC (Colder) PMCD1604 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/4" ID, 1/8" Flow, Acetal
Buy More Save More
EW-06360-15 Mfr # PMCD1604
COMPARE
1/4 Acetal Quick-Disconnect x Barb, Valved
$15.70USD / Each
CPC (Colder) PMC1601 Quick-Disconnect Panel-Mount Hose Barb Body; 1/16" ID, 1/8" Flow, Acetal
Buy More Save More
EW-06361-03 Mfr # PMC1601
COMPARE
1/16 Acetal Quick-Disconnect x Barb, Nonvalved
$13.60USD / Each
CPC (Colder) PMC1602 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 1/8" Tube ID, 1/8" Flow Size
Buy More Save More
EW-06361-04 Mfr # PMC1602
COMPARE
1/8 Acetal Quick-Disconnect x Barb, Nonvalved
$14.10USD / Each
CPC (Colder) PMC1603 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 3/16" Tube ID, 1/8" Flow Size
Buy More Save More
EW-06361-05 Mfr # PMC1603
COMPARE
3/16 Acetal Quick-Disconnect x Barb, Nonvalved
$14.00USD / Each
CPC (Colder) PMCD160112 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/16" ID, 1/8" Flow, Acetal
Buy More Save More
EW-06364-09 Mfr # PMCD160112
COMPARE
1/16 Polypropylene Quick-Disconnect x Barb, Valved
$17.00USD / Each
CPC (Colder) PMCD160212 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 1/8" Tube ID, PP
Buy More Save More
EW-06364-10 Mfr # PMCD160212
COMPARE
1/8 Polypropylene Quick-Disconnect x Barb, Valved
$16.30USD / Each
CPC (Colder) PMCD160412 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/4" ID, 1/8" Flow, PP
Buy More Save More
EW-06364-15 Mfr # PMCD160412
COMPARE
1/4 Polypropylene Quick-Disconnect x Barb, Valved
$16.30USD / Each
CPC (Colder) PMC1604 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 1/4" Tube ID, 1/8" Flow Size; 25/PK EW-31307-04 Mfr # PMC1604
COMPARE
1/4 Acetal Quick-Disconnect x Barb, Nonvalved
$301.00USD / Pkg of 25
Showing 1- 11 of 11