Parafilm® M Wraps
Colorless, moisture proof plastic film molds to most surfaces
  • Prevents moisture and volume loss
  • Reduces risk of tampered data
  • Clings around irregular shapes and surfaces
  • Thermoplastic film with two-way stretch
The Parafilm® M wrap is a colorless, moisture-proof film that molds to the surface. It is a semi-transparent, flexible, thermoplastic film with two-way stretch. It is resistant up to 48 hours against many polar substances and prevents moisture and volume loss due to the material composition and tight seal. Ideal accessory for any laboratory. Helps protect the integrity of your laboratory and research applications.
$41.50 - $110.00USD / Roll of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (ft) Width (in)
Availability Pricing
Parafilm PM998 M Wrap, 20" Wide; 50 Ft/Roll
Always in Stock
EW-06720-25 Mfr # PM998
COMPARE
Semi-transparent 50 20
$110.00USD / Roll of 1
Parafilm PM996 M Wrap, 4" Wide; 125 Ft/Roll
Always in Stock
EW-06720-40 Mfr # PM996
COMPARE
Semi-transparent 125 4
$45.00USD / Roll of 1
Parafilm PM992 M Wrap, 2" Wide; 250 Ft/Roll
Always in Stock
EW-06720-50 Mfr # PM992
COMPARE
Semi-transparent 250 2
$41.50USD / Roll of 1
Parafilm PM999 M Wrap, 4" Wide; 250Ft/Roll
Always in Stock
EW-44002-46 Mfr # HS234526C
COMPARE
Natural 250 4
$75.00USD / Each