Parker Hannifin Adapter Fittings, Compression to Male Threaded, Elbow
Excellent choice to connect rigid tubing systems
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression and threaded fittings offer a more permanent installation. Choose the size and material that suit your application needs.
$37.75 - $178.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
Parker Hannifin Fitting, Black Polypropylene, Elbow, Compression to Threaded Adapter, 1/4" OD x 1/8" NPT(M); 25/PK
Buy More Save More
EW-06444-20 Mfr # P4ME2
COMPARE
1/4 Polypropylene 1/8
$104.00USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Elbow, Compression to Threaded Adapter, 1/4" OD x 1/4" NPT(M); 25/PK
Buy More Save More
EW-06444-21 Mfr # P4ME4
COMPARE
1/4 Polypropylene 1/4
$85.50USD / of 25
Parker Hannifin P6ME6 Fitting, Black Polypropylene, Elbow, Compression to Threaded Adapter, 3/8"OD x 3/8" NPT(M); 25/PK
Buy More Save More
EW-06444-22 Mfr # P6ME6
COMPARE
3/8 Polypropylene 3/8
$118.00USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Elbow, Compression to Threaded Adapter, 3/8" OD x 1/2" NPT(M); 25/PK
Buy More Save More
EW-06444-23 Mfr # P6ME8
COMPARE
3/8 Polypropylene 1/2
$134.00USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Elbow, Compression to Threaded Adapter, 1/2" OD x 1/2" NPT(M); 25/PK
Buy More Save More
EW-06444-24 Mfr # P8ME8
COMPARE
1/2 Polypropylene 1/2
$167.00USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Elbow, Compression to Threaded Adapter, 5/8" OD x 1/2" NPT(M); 25/PK
Buy More Save More
EW-06444-25 Mfr # P10ME8
COMPARE
5/8 Polypropylene 1/2
$178.00USD / of 25
Parker Hannifin Fitting, Stainless Steel, Elbow, Compression to Threaded Adapter, 1/8" OD x 1/8" NPT(M); 1/EA EW-30486-52 Mfr # 2MSELN2N-316
COMPARE
1/8 Stainless Steel 1/8
$41.75USD / Each
Parker Hannifin 2MSELN4N-316 Fitting, Stainless Steel, Elbow, Compression to Threaded Adapter, 1/8" OD x 1/4" NPT(M); 1/EA EW-30486-53 Mfr # 2MSELN4N-316
COMPARE
1/8 Stainless Steel 1/4
$48.00USD / Each
Parker Hannifin 4MSELN2N-316 Fitting, Stainless Steel, Elbow, Compression to Threaded Adapter, 1/4" OD x 1/8" NPT(M); 1/EA EW-30486-54 Mfr # 4MSELN2N-316
COMPARE
1/4 Stainless Steel 1/8
$37.75USD / Each
Parker Hannifin Fitting, Stainless Steel, Elbow, Compression to Threaded Adapter, 1/4" OD x 1/4" NPT(M); 1/EA EW-30486-55 Mfr # 4MSELN4N-316
COMPARE
1/4 Stainless Steel 1/4
$41.75USD / Each
Parker Hannifin Fitting, Stainless Steel, Elbow, Compression to Threaded Adapter, 3/8" OD x 1/4" NPT(M); 1/EA EW-30486-56 Mfr # 6MSEL4N-316
COMPARE
3/8 Stainless Steel 1/4
$49.50USD / Each
Parker Hannifin Fitting, Stainless Steel, Elbow, Compression to Threaded Adapter, 3/8" OD x 3/8" NPT(M); 1/EA EW-30486-57 Mfr # 6MSELN6N-316
COMPARE
3/8 Stainless Steel 3/8
$56.00USD / Each
Showing 1- 12 of 12