Parker Hannifin® Flare Union Elbows, PFA
Leak-tight and high-pressure connection for corrosive chemical applications
  • Constructed of durable PFA
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.
$82.50 - $156.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Fitting Material
Availability Pricing
Parker Hannifin Flare Union Fitting, Elbow, 1/4", PFA; 1/Ea EW-30490-46 Mfr # FEC-4
COMPARE
1/4 1/4 PFA
$82.50USD / Each
Parker Hannifin Flare Union Fitting, Elbow, 3/8", PFA; 1/Ea EW-30490-48 Mfr # FEC-6
COMPARE
3/8 3/8 PFA
$100.00USD / Each
Parker Hannifin FEC-8 Flare Union Fitting, Elbow, 1/2", PFA; 1/Ea EW-30490-50 Mfr # FEC-8
COMPARE
1/2 1/2 PFA
$115.50USD / Each
Parker Hannifin FEC-12 Flare Union Fitting, Elbow, 3/4", PFA; 1/Ea EW-30490-52 Mfr # FEC-12
COMPARE
3/4 3/4 PFA
$156.50USD / Each