Parker Hannifin Metal Two-way Right-Angled Ball Valves
Metal construction for druable applications
  • Constructed of metal parts for durable applications
Ball valves are designed primarily for on/off service. The two-way valve model has a single inlet port and a single outlet port for straight through operation. The handle of the valve can be turned to change the flow pattern or on/off service of the valve.
$153.25 - $238.75USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
Parker Hannifin Two-way right angled ball valve, Stainless Steel, 1/4" compression connection EW-30527-48 Mfr # 4A-MB4APFA-SSP
COMPARE
1/4" compression 1/4" compression Stainless Steel, PFA
$168.00USD / Each
Parker Hannifin two-way right angled ball valve, stainless steel, 1/4" NPT(F) EW-30527-50 Mfr # 4F-MB6APFA-SSP
COMPARE
1/4" NPT(F) 1/4" NPT(F) Stainless Steel, PFA
$192.00USD / Each
Parker Hannifin two-way right angled ball valve, stainless steel, 3/8" compression connection EW-30527-52 Mfr # 6A-MB6APFA-SSP
COMPARE
3/8" compression 3/8" compression Stainless Steel, PFA
$238.75USD / Each
Parker Hannifin two-way right angled ball valve, brass, 1/4" NPT(F) EW-30527-76 Mfr # 4F-MB6APFA-BP
COMPARE
1/4" NPT(F) 1/4" NPT(F) Brass, PFA
$153.25USD / Each