Parker Hannifin Poppet Check Valves, PFA
Install in any position
  • Constructed of chemically inert PFA with Viton O-rings
Valves have body made of PFA and O-ring seals made of Viton®. Max pressure is 100 psi; temperature range is 0 to 300°F (–18 to 149°C). Install in any position.
Note: Valves require 5 psi back pressure to achieve a proper seal.
$345.00 - $433.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Breaking Pressure (PSI) Cv Port Size (NPT(F))
Availability Pricing
Parker Hannifin CV-1-1144 Poppet Check Valve, 1/4" NPT(F) connections EW-06373-57 Mfr # CV-1-1144
COMPARE
1 0.75 0.25
$345.00USD / Each
Parker Hannifin CV-1-1188 Poppet Check Valve, 1/2" NPT(F) connections EW-06373-59 Mfr # CV-1-1188
COMPARE
1.75 4 0.5
$433.00USD / Each