Parker Hannifin Push-to-Connect Check Valves, Acetal
Create a secure connection with your check valve
  • Constructed of durable acetal plastic
Use these push-to-connect check valves for rigid and semirigid plastic tubing. Acetal body with EPDM O-rings. Temperature range of 34°F (1.1°C) to 150°F (65.6°C).
$19.30 - $22.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Breaking Pressure (PSI) Inlet Port Outlet Port
Availability Pricing
Parker Hannifin CHECK VALVE, ACETAL BODY, push-to-connect, 1/4'' tube OD EW-01379-89 Mfr # A4VC4-MG
COMPARE
0.333 1/4" Push to Connect 1/4" Push to Connect
$19.50USD / Each
Parker Hannifin CHECK VALVE, ACETAL BODY, push-to connect, 5/16'' tube OD EW-01379-91 Mfr # A5VC5-MG
COMPARE
0.333 5/16" Push to Connect 5/16" Push to Connect
$19.30USD / Each
Parker Hannifin Check Valve, Acetal Body, push-to-connect, 3/8'' tube OD EW-01379-93 Mfr # A6VC6-MG
COMPARE
0.333 3/8" Push to Connect 3/8" Push to Connect
$22.50USD / Each