Parker Hannifin Union Fittings, Compression, Straight
Get a secure seal at higher pressures
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
$29.25 - $182.00USD / of 25
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Fitting Material
Availability Pricing
Parker Hannifin Fitting, Black Polypropylene, Straight, Compression Union, 1/4" OD; 25/PK
Buy More Save More
EW-06444-51 Mfr # P4UC4
COMPARE
1/4 1/4 Polypropylene
$99.50USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Straight, Compression Union, 3/8" OD; 25/PK
Buy More Save More
EW-06444-52 Mfr # P6UC6
COMPARE
3/8 3/8 Polypropylene
$131.00USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Straight, Compression Union, 1/2" OD; 25/PK
Buy More Save More
EW-06444-54 Mfr # P8UC8
COMPARE
1/2 1/2 Polypropylene
$160.00USD / of 25
Parker Hannifin Fitting, Black Polypropylene, Straight, Compression Union, 5/8" OD; 25/PK
Buy More Save More
EW-06444-56 Mfr # P10UC10
COMPARE
5/8 5/8 Polypropylene
$182.00USD / of 25
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 1/8" OD
Buy More Save More
EW-30486-00 Mfr # 2SC2-316
COMPARE
1/8 1/8 316 Stainless Steel
$29.25USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 1/4" OD
Buy More Save More
EW-30486-01 Mfr # 4SC4-316
COMPARE
1/4 1/4 316 Stainless Steel
$41.25USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 5/16" OD
Buy More Save More
EW-30486-02 Mfr # 5SC5-316
COMPARE
5/16 5/16 316 Stainless Steel
$33.75USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 3/8" OD
Buy More Save More
EW-30486-03 Mfr # 6SC6-316
COMPARE
3/8 3/8 316 Stainless Steel
$53.00USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 1/2" OD
Buy More Save More
EW-30486-04 Mfr # 8SC8-316
COMPARE
1/2 1/2 316 Stainless Steel
$75.00USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 5/8" OD"
Buy More Save More
EW-30486-05 Mfr # 10SC10-316
COMPARE
1/16 5/8 316 Stainless Steel
$83.00USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 3/4" OD"
Buy More Save More
EW-30486-06 Mfr # 12SC12-316
COMPARE
3/4 3/4 316 Stainless Steel
$95.00USD / Each
Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 1/16" OD
Buy More Save More
EW-30486-70 Mfr # 1-1 HBZ-SS
COMPARE
1/16 1/16 316 Stainless Steel
$42.25USD / Each
Parker Hannifin Fitting, PFA, Straight, Compression Union,  1/8" OD
Buy More Save More
EW-31320-21 Mfr # GSC-2
COMPARE
1/8 1/8 PFA
$47.25USD / Each
Parker Hannifin Fitting, PFA, Straight, Compression Union, 1/4" OD
Buy More Save More
EW-31320-22 Mfr # GSC-4
COMPARE
1/4 1/4 PFA
$53.00USD / Each
Parker Hannifin Fitting, PFA, Straight, Compression Union, 3/8" OD
Buy More Save More
EW-31320-23 Mfr # GSC-6
COMPARE
3/8 3/8 PFA
$57.50USD / Each
Parker Hannifin Fitting, PFA, Straight, Compression Union, 3/4" OD
Buy More Save More
EW-31320-24 Mfr # GSC-12
COMPARE
3/4 3/4 PFA
$96.00USD / Each
COMPARE
5/32 5/32 PFA
$73.00USD / Each
COMPARE
15/64 15/64 PFA
$69.00USD / Each
Parker Hannifin Fitting, PFA, Straight, Compression Union, 1/2" OD
Buy More Save More
EW-31321-65 Mfr # GSC-8
COMPARE
1/2 1/2 PFA
$55.50USD / Each
Showing 1- 19 of 19