Parker Hannifin XM Air-Control Valves
  • Continuous duty rated
  • Direct solenoid, spring return
  • No minimum operating pressure required — solenoid coil operates an armature pin that directly impacts the spool mechanism to shift the valve
Although there is no minimum operating pressure, the user of the XM valve must choose the valve for the system pressure. A 3-way valve that is used in vacuum service is constructed differently from a valve that is used in positive pressure systems. Four-way valves can be used in vacuum or positive pressure systems.
The XM valve is a non-repairable valve. Once the stroke is set on the solenoid armature, the valve is set. This makes the internal mechanism of the XM valve non-repairable.
All Ports 1/8. The inline valves can be mounted as a single valve. Valve has mounting holes. Inline valves can be manifold mounted on an IEM manifold. Manifold accepts both 3-way and 4-way valves without any type of additional kits. IEM manifold ports are 1/4.
$13.00 - $210.00USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description Flow Pattern Max Pressure (PSI)
Availability Pricing
Parker Hannifin XM30NBG53A 2-Position, 3-Way Body Ported Valve; 0-120 psig; 120VAC EW-98623-01 Mfr # XM30NBG53A
COMPARE
2-Position, 3-Way Body Ported Valve; 0-120 psig; 120VAC Three-Way, Two-Position 120
$106.00USD / Each
Parker Hannifin XM30NBG49A 2-Position, 3-Way Body Ported Valve; 0-120 psig; 24VDC EW-98623-03 Mfr # XM30NBG49A
COMPARE
2-Position, 3-Way Body Ported Valve; 0-120 psig; 24VDC Three-Way, Two-Position 120
$79.00USD / Each
Parker Hannifin XM30NBG45A 2-Position, 3-Way Body Ported Valve; 0-120 psig; 12VDC
Always in Stock
EW-98623-05 Mfr # XM30NBG45A
COMPARE
2-Position, 3-Way Body Ported Valve; 0-120 psig; 12VDC Three-Way, Two-Position 120
$98.50USD / Each
Parker Hannifin XM30VBG53A 2-Position, 3-Way Body Ported Valve; -14.7 to 120 psig; 120VAC EW-98623-07 Mfr # XM30VBG53A
COMPARE
2-Position, 3-Way Body Ported Valve; -14.7 to 120 psig; 120VAC Three-Way, Two-Position 15
$106.00USD / Each
Parker Hannifin XM30VBG49A 2-Position, 3-Way Body Ported Valve; -14.7 to 15psig; 24VDC EW-98623-09 Mfr # XM30VBG49A
COMPARE
2-Position, 3-Way Body Ported Valve; -14.7 to 15psig; 24VDC
$96.00USD / Each
Parker Hannifin XM30VBG45A 2-Position, 3-Way Body Ported Valve; -14.7 to 15psig; 12VDC EW-98623-11 Mfr # XM30VBG45A
COMPARE
2-Position, 3-Way Body Ported Valve; -14.7 to 15psig; 12VDC
$96.00USD / Each
Parker Hannifin XM40NBG53A 2-Position, 4-Way Body Ported Valve; -14.7 to 125psig; 120VAC EW-98623-13 Mfr # XM40NBG53A
COMPARE
2-Position, 4-Way Body Ported Valve; -14.7 to 125psig; 120VAC
$159.00USD / Each
Parker Hannifin XM40NBG49A 2-Position, 4-Way Body Ported Valve; -14.7 to 125psig; 24VDC EW-98623-15 Mfr # XM40NBG49A
COMPARE
2-Position, 4-Way Body Ported Valve; -14.7 to 125psig; 24VDC
$153.00USD / Each
Parker Hannifin XM40NBG45A 2-Position, 4-Way Body Ported Valve; -14.7 to 125psig; 12VDC EW-98623-17 Mfr # XM40NBG45A
COMPARE
2-Position, 4-Way Body Ported Valve; -14.7 to 125psig; 12VDC
$153.00USD / Each
Parker Hannifin PSMXNXN02NP 2-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports EW-98623-30 Mfr # PSMXNXN02NP
COMPARE
2-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports
$71.50USD / Each
Parker Hannifin PSMXNXN04NP 4-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports EW-98623-31 Mfr # PSMXNXN04NP
COMPARE
4-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports
$123.00USD / Each
Parker Hannifin PSMXNXN06NP 6-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports EW-98623-32 Mfr # PSMXNXN06NP
COMPARE
6-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports
$150.00USD / Each
Parker Hannifin PSMXNXN08NP 8-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports EW-98623-33 Mfr # PSMXNXN08NP
COMPARE
8-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports
$179.00USD / Each
Parker Hannifin PSMXNXN10NP 10-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports EW-98623-34 Mfr # PSMXNXN10NP
COMPARE
10-Station Manifold for XM Solenoid Valves, 1/4" NPT(F) ports
$210.00USD / Each
Parker Hannifin PSXM8288P Inline Valve Mounting Foot Bracket for XM Solenoid Valves EW-98623-41 Mfr # PSXM8288P
COMPARE
Inline Valve Mounting Foot Bracket for XM Solenoid Valves
$13.00USD / Each
Parker Hannifin EM12 Muffler for XM Solenoid Valves, 1/8" NPT(M), sintered bronze EW-98623-42 Mfr # EM12
COMPARE
Muffler for XM Solenoid Valves, 1/8" NPT(M), sintered bronze
$15.75USD / Each
Parker Hannifin EM25 Muffler for XM Solenoid Valves, 1/4" NPT(M), sintered bronze EW-98623-45 Mfr # EM25
COMPARE
Muffler for XM Solenoid Valves, 1/4" NPT(M), sintered bronze
$17.00USD / Each
Parker Hannifin PSXM8310P Valve Station Blanking Plate for XM Solenoid Valve Manifolds EW-98623-46 Mfr # PSXM8310P
COMPARE
Valve Station Blanking Plate for XM Solenoid Valve Manifolds
$26.50USD / Each
Showing 1- 18 of 18
Need Help with this product?
Call Us At1-847-549-7600
Live chat