Parker High-Pressure Metal Three-way Ball Valves
Don’t worry about leaks
Ball valves are designed primarily for on/off service. The three-way valve model has three ports, and a combination of the ports can be either open or closed for various flow pattern capabilities. The handle of the valve can be turned to change the flow pattern or on/off service of the valve. Choose from stainless steel/PFA or brass/PFA valve options.
$99.50 - $222.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
Parker Hannifin 2A-MB2XPFA-SSP three-way ball valve, stainless steel, 1/8" compression connection EW-30527-54 Mfr # 2A-MB2XPFA-SSP
COMPARE
1/8" compression 1/8" compression Stainless Steel
$158.00USD / Each
Parker Hannifin 2F-MB4XPFA-SSP three-way ball valve, stainless steel, 1/8" NPT(F) EW-30527-56 Mfr # 2F-MB4XPFA-SSP
COMPARE
1/8" NPT(F) 1/8" NPT(F) Stainless Steel
$144.00USD / Each
Parker Hannifin 4A-MB4XPFA-SSP three-way ball valve, stainless steel, 1/4" compression connection EW-30527-58 Mfr # 4A-MB4XPFA-SSP
COMPARE
1/4" compression 1/4" 压缩 Stainless Steel
$160.00USD / Each
Parker Hannifin 4F-MB6XPFA-SSP three-way ball valve, stainless steel, 1/4" NPT(F) EW-30527-60 Mfr # 4F-MB6XPFA-SSP
COMPARE
1/4" NPT(F) 1/4" NPT(F) Stainless Steel
$176.00USD / Each
Parker Hannifin 6A-MB6XPFA-SSP three-way ball valve, stainless steel, 3/8" compression connection EW-30527-62 Mfr # 6A-MB6XPFA-SSP
COMPARE
3/8" compression 3/8" compression Stainless Steel
$222.00USD / Each
Parker Hannifin 2A-MB2XPFA-BP three-way ball valve, brass, 1/8" compression connection EW-30527-80 Mfr # 2A-MB2XPFA-BP
COMPARE
1/8" compression 1/8" compression Brass
$99.50USD / Each
Parker Hannifin 2F-MB4XPFA-BP three-way ball valve, brass, 1/8" NPT(F) EW-30527-82 Mfr # 2F-MB4XPFA-BP
COMPARE
1/8" NPT(F) 1/8" NPT(F) Brass
$99.50USD / Each
Parker Hannifin 4A-MB4XPFA-BP three-way ball valve, brass, 1/4" compression connection EW-30527-84 Mfr # 4A-MB4XPFA-BP
COMPARE
1/4" compression 1/4" compression Brass
$99.50USD / Each
Parker Hannifin 4F-MB6XPFA-BP three-way ball valve, brass, 1/4" NPT(F) EW-30527-86 Mfr # 4F-MB6XPFA-BP
COMPARE
1/4" NPT(F) 1/4" NPT(F) Brass
$115.00USD / Each
Parker Hannifin 6A-MB6XPFA-BP three-way ball valve, brass, 3/8" compression connection EW-30527-88 Mfr # 6A-MB6XPFA-BP
COMPARE
3/8" compression 3/8" compression Brass
$140.00USD / Each
Showing 1- 10 of 10