Parker High-Pressure Metal Two-way Ball Valves (Straight)
Don’t worry about leaks
Ball valves are designed primarily for on/off service. The two-way valve model has a single inlet port and a single outlet port for straight through operation. The handle of the valve can be turned to change the flow pattern or on/off service of the valve. Choose from stainless steel/PFA or brass/PFA valve options.
$83.00 - $376.50USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Orifice (in) Outlet Port
Availability Pricing
Parker Hannifin 2A-MB2LPFA-SSP two-way straight ball valve, stainless steel, 1/8" compression connection EW-30527-30 Mfr # 2A-MB2LPFA-SSP
COMPARE
1/8" compression 0.093 1/8" compression
$139.50USD / Each
Parker Hannifin 2F-MB4LPFA-SSP two-way straight ball valve, stainless steel, 1/8" NPT(F) EW-30527-32 Mfr # 2F-MB4LPFA-SSP
COMPARE
1/8" NPT(F) 0.125 1/8" NPT(F)
$138.00USD / Each
Parker Hannifin 4A-MB4LPFA-SSP two-way straight ball valve, stainless steel, 1/4" compression connection EW-30527-34 Mfr # 4A-MB4LPFA-SSP
COMPARE
1/4" compression 0.125 1/4" compression
$142.00USD / Each
Parker Hannifin 4F-MB6LPFA-SSP two-way straight ball valve, stainless steel, 1/4" NPT(F) EW-30527-36 Mfr # 4F-MB6LPFA-SSP
COMPARE
1/4" NPT(F) 0.187 1/4" NPT(F)
$148.00USD / Each
Parker Hannifin 4M-MB6LPFA-SSP two-way straight ball valve, stainless steel, 1/4" NPT(M) EW-30527-38 Mfr # 4M-MB6LPFA-SSP
COMPARE
1/4" NPT(M) 0.187 1/4" NPT(M)
$158.50USD / Each
Parker Hannifin 6A-MB6LPFA-SSP two-way straight ball valve, stainless steel, 3/8" compression connection EW-30527-40 Mfr # 6A-MB6LPFA-SSP
COMPARE
3/8" compression 0.187 3/8" compression
$158.50USD / Each
Parker Hannifin 8A-MB8LPFA-SSP two-way straight ball valve, stainless steel, 1/2" compression connection EW-30527-42 Mfr # 8A-MB8LPFA-SSP
COMPARE
1/2" compression 0.41 1/2" compression
$376.50USD / Each
Parker Hannifin 8F-MB8LPFA-SSP two-way straight ball valve, stainless steel, 1/2" NPT(F) EW-30527-44 Mfr # 8F-MB8LPFA-SSP
COMPARE
1/2" NPT(F) 0.41 1/2" NPT(F)
$368.50USD / Each
Parker Hannifin 2A-MB2LPFA-BP two-way straight ball valve, brass, 1/8" compression connection EW-30527-64 Mfr # 2A-MB2LPFA-BP
COMPARE
1/8" compression 0.093 1/8" compression
$85.50USD / Each
Parker Hannifin 2F-MB4LPFA-BP two-way straight ball valve, brass, 1/8" NPT(F) EW-30527-66 Mfr # 2F-MB4LPFA-BP
COMPARE
1/8" NPT(F) 0.125 1/8" NPT(F)
$83.00USD / Each
Parker Hannifin 4A-MB4LPFA-BP two-way straight ball valve, brass, 1/4" compression connection EW-30527-68 Mfr # 4A-MB4LPFA-BP
COMPARE
1/4" compression 0.125 1/4" compression
$87.50USD / Each
Parker Hannifin 4F-MB6LPFA-BP two-way straight ball valve, brass, 1/4" NPT(F) EW-30527-70 Mfr # 4F-MB6LPFA-BP
COMPARE
1/4" NPT(F) 0.187 1/4" NPT(F)
$103.50USD / Each
Parker Hannifin 6A-MB6LPFA-BP two-way straight ball valve, brass, 3/8" compression connection EW-30527-74 Mfr # 6A-MB6LPFA-BP
COMPARE
3/8" compression 0.187 3/8" compression
$109.00USD / Each
Showing 1- 13 of 13