Parker Needle Valves
  • Valves are designed for positive leak tight shut-off and regulation of fluids in process, power, and instrumentation applications
  • All metal tapered needle stem tip
  • Differential hardness between the strain hardened steam and cold formed body threads provides improved cycle life
  • Panel mountable — order panel-mounting nuts separately
$81.50 - $228.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate For Air (LPM) Tubing OD (in) Wetted Materials
Availability Pricing
Parker Hannifin 2A-V2LN-SS needle valve; 1/8" compression connectors, 316 SS EW-06394-05 Mfr # 2A-V2LN-SS
COMPARE
10000 0.125 Stainless Steel
$158.00USD / Each
Parker Hannifin 4A-V4LN-SS needle valve; 1/4" compression connectors, 316 SS EW-06394-07 Mfr # 4A-V4LN-SS
COMPARE
36000 0.25 Stainless Steel
$139.00USD / Each
Parker Hannifin 6A-V6LN-SS needle valve; 3/8" compression connectors, 316 SS EW-06394-09 Mfr # 6A-V6LN-SS
COMPARE
60000 0.375 Stainless Steel
$183.00USD / Each
Parker Hannifin 8A-V8LN-SS needle valve; 1/2" compression connectors, 316 SS EW-06394-11 Mfr # 8A-V8LN-SS
COMPARE
10000 0.5 Stainless Steel
$228.00USD / Each
Parker Hannifin 4A-V4LN-B needle valve; 1/4" compression connectors, brass EW-06394-13 Mfr # 4A-V4LN-B
COMPARE
12500 0.25 Brass
$81.50USD / Each
Parker Hannifin 6A-V6LN-B needle valve; 3/8" compression connectors, brass EW-06394-15 Mfr # 6A-V6LN-B
COMPARE
35000 0.375 Brass
$100.00USD / Each
Parker Hannifin 8A-V8LN-B needle valve; 1/2" compression connectors, brass EW-06394-17 Mfr # 8A-V8LN-B
COMPARE
62000 0.5 Brass
$169.00USD / Each
Showing 1- 7 of 7
Need Help with this product?
Call Us At1-847-549-7600
Live chat