Parker Polypropylene Ball Valves
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
  • All plastic design is leak free and corrosion resistant
$10.90 - $24.75USD / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Orifice (in) Outlet Port
Availability Pricing
Parker Hannifin PP4VME4MG Valve, Ball, Polypropylene, Male Elbow 1/4'' push to connect tube, 1/4'' NPT(M) EW-01379-41 Mfr # PP4VME4-MG
COMPARE
1/4" NPT(M) 0.1875 1/4" Push to Connect
$13.50USD / Each
Parker Hannifin PP4VME6-MG Valve, Ball, Polypropylene, Male Elbow 1/4'' push to connect tube, 3/8'' NPT(M) EW-01379-43 Mfr # PP4VME6-MG
COMPARE
3/8" NPT(M) 0.1875 1/4" Push to Connect
$18.20USD / Each
Parker Hannifin PP6VME4-MG Ball Valve, Polypropylene, Male Elbow 3/8'' push-to-connect tube, 1/4'' NPT(M) EW-01379-47 Mfr # PP6VME4-MG
COMPARE
1/4" NPT(M) 0.25 3/8" Push to Connect
$12.90USD / Each
Parker Hannifin PP6VME8-MG Valve, Ball, Polypropylene, Male Elbow 3/8'' push-to-connect tube, 1/2'' NPT(M) EW-01379-51 Mfr # PP6VME8-MG
COMPARE
1/2" NPT(M) 0.25 3/8" Push to Connect
$21.25USD / Each
Parker Hannifin PP4VFE6-MG Valve, Ball, Polypropylene, Female Elbow 1/4'' push to connect tube, 3/8'' NPT(F) EW-01379-55 Mfr # PP4VFE6-MG
COMPARE
3/8" NPT(F) 0.1875 1/4" Push to Connect
$14.30USD / Each
Parker Hannifin PP6VFE4-MG Ball Valve, Polypropylene, Female Elbow 3/8'' push to connect tube, 1/4'' NPT(F) EW-01379-57 Mfr # PP6VFE4-MG
COMPARE
1/4" NPT(F) 0.25 3/8" Push to Connect
$12.20USD / Each
Parker Hannifin PP6VFE6-MG Ball Valve, Polypropylene, Female Elbow 3/8'' push to connect x 3/8 NPT tube EW-01379-59 Mfr # PP6VFE6-MG
COMPARE
3/8" NPT(F) 0.25 3/8" Push to Connect
$12.40USD / Each
Parker Hannifin PP4VUC6-MG Ball Valve, Polypropylene, Male Elbow 1/4'', 3/8'' push to connect tube EW-01379-63 Mfr # PP4VUC6-MG
COMPARE
1/4" Push to Connect 0.1875 3/8" Push to Connect
$16.90USD / Each
Parker Hannifin PP6VUC4-MG Ball Valve, Polypropylene, Male Elbow 3/8'', 1/4'' push to connect tube, 1/each EW-01379-65 Mfr # PP6VUC4-MG
COMPARE
3/8" Push to Connect 0.25 1/4" Push to Connect
$16.40USD / Each
Parker Hannifin PP4VMC6-MG Ball Valve, Polypropylene, Male Connector 1/4'' push-to-connect tube, 3/8'' NPT(M) EW-01379-71 Mfr # PP4VMC6-MG
COMPARE
3/8" NPT(M) 0.1875 1/4" Push to Connect
$13.80USD / Each
Parker Hannifin PP6VMC4-MG Ball Valve, Polypropylene, Male Connector 3/8'' push-to-connect tube, 1/4'' NPT(M) EW-01379-73 Mfr # PP6VMC4-MG
COMPARE
1/4" NPT(M) 0.25 3/8" Push to Connect
$12.90USD / Each
Parker Hannifin PP6VMC6-MG Valve, Ball, Polypropylene, Male Connector 3/8'' push-to-onnect tube, 3/8'' NPT(M) EW-01379-75 Mfr # PP6VMC6-MG
COMPARE
3/8" NPT(M) 0.25 3/8" Push to Connect
$11.40USD / Each
Parker Hannifin PP4VFC2MG Ball Valve, Polypropylene, Female Connector 1/4'' push-to-connect tube, 1/8'' NPT(F) EW-01379-77 Mfr # PP4VFC2-MG
COMPARE
1/8" NPT(F) 0.1875 1/4" Push to Connect
$14.30USD / Each
Parker Hannifin PP4VFC4MG Ball Valve, Polypropylene, Female Connector 1/4'' push-to-connect tube, 1/4'' NPT(F) EW-01379-79 Mfr # PP4VFC4-MG
COMPARE
1/4" NPT(F) 0.1875 1/4" Push to Connect
$14.10USD / Each
Parker Hannifin PP4VFC6MG Ball Valve, Polypropylene, Female Connector 1/4'' push-to-connect tube, 3/8'' NPT(F) EW-01379-81 Mfr # PP4VFC6-MG
COMPARE
3/8" NPT(F) 0.1875 1/4" Push to Connect
$13.80USD / Each
Parker Hannifin PP6VFC2-MG Ball Valve, Polypropylene, Female Connector 3/8'' push-to-connect tube, 1/8'' NPT(F) EW-01379-83 Mfr # PP6VFC2-MG
COMPARE
1/8" NPT(F) 0.25 3/8" Push to Connect
$16.60USD / Each
Parker Hannifin PP6VFC4-MG Ball Valve, Polypropylene, Female Connector 3/8'' push-to-connect tube, 1/4'' NPT(F) EW-01379-85 Mfr # PP6VFC4-MG
COMPARE
1/4" NPT(F) 0.25 3/8" Push to Connect
$15.90USD / Each
Parker Hannifin PP6VFC6-MG Ball Valve, Polypropylene, Female Connector 3/8'' push-to-connect tube, 3/8'' NPT(F) EW-01379-87 Mfr # PP6VFC6-MG
COMPARE
3/8" NPT(F) 0.25 3/8" Push to Connect
$13.80USD / Each
Parker Hannifin PP4VTEU6-MG Valve, Ball, Polypropylene, Tube Elbow 1/4'' push-to-connect tube, 3/8'' stem, 1/each EW-01379-95 Mfr # PP4VTEU6-MG
COMPARE
3/8" stem 0.25 1/4" Push to Connect
$18.50USD / Each
Parker Hannifin PP6VTEU6-MG Valve, Ball, Polypropylene, Tube Elbow 3/8'' push-to-connect tube, 3/8'' stem, 1/each EW-01379-97 Mfr # PP6VTEU6-MG
COMPARE
3/8" stem 0.375 3/8" Push to Connect
$24.75USD / Each
Showing 1 - 20 of 26