Particle Tolerant Solenoid Valves
  • Special elastomer diaphragm and seals are forgiving of particulate matter that would typically damage a plastic valve seat
  • Inert materials — excellent chemical resistance
  • Molded PEEK valve bodies
  • Excellent choice for demanding low-flow applications where minimum size and internal volume are important
$164.00 - $360.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Max Pressure (PSI) Orifice (in)
Availability Pricing
COMPARE
1/4"-28 UNF 0.0472
$164.00USD / Each
COMPARE
1/4"-28 UNF Inlet: 29 Outlet: 7 0.0472
$164.00USD / Each
COMPARE
1/4"-28 UNF 0.0787
$192.00USD / Each
COMPARE
1/4"-28 UNF Inlet: 29 Outlet: 14 0.0787
$192.00USD / Each
COMPARE
1/4"-28 UNF 0.0472
$237.00USD / Each
COMPARE
1/4"-28 UNF Inlet: 29 Outlet: 7 0.0472
$234.00USD / Each
COMPARE
1/4"-28 UNF 0.0787
$297.00USD / Each
COMPARE
1/4"-28 UNF Common: 29; NC/NO: 7 0.0472
$248.00USD / Each
COMPARE
1/4"-28 UNF Inlet: 29 Outlet: 14 0.0787
$297.00USD / Each
COMPARE
1/4"-28 UNF Common: 29; NC/NO: 14 0.0787
$360.00USD / Each
COMPARE
1/4"-28 UNF Common: 29; NC/NO: 14 0.0787
$358.00USD / Each
Showing 1- 11 of 11
Need Help with this product?
Call Us At1-847-549-7600
Live chat