This product is now discontinued, View Related Products
PCRmax Colored Polypropylene PCR Tubes
Obtain short cycle times while staying organized
  • Integrated flip-cap, matte writing surface, and five colors to choose from
  • Ultra-thin walls for rapid heat transfer and faster cycle times
  • High-purity polypropylene construction is free from RNase, DNase, endotoxins, and human DNA
  • Withstand temperatures of –20 to 122°C
PCRmax consumable products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.
Autoclavable
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Color Description
Availability Pricing
COMPARE
0.2 Clear Colored PCR Tubes, Clear, 0.2 mL; 500/Pk
Call For Price
COMPARE
0.2 Red Colored PCR Tubes, Red, 0.2 mL; 500/Pk
Call For Price
COMPARE
0.2 Clear Colored PCR Tubes, Yellow, 0.2 mL; 500/Pk
Call For Price
COMPARE
0.2 Clear Colored PCR Tubes, Green, 0.2 mL; 500/Pk
Call For Price
COMPARE
0.2 Clear Colored PCR Tubes, Blue, 0.2 mL; 500/Pk
Call For Price
COMPARE
0.5 Clear Colored PCR Tubes, Clear, 0.5 mL; 1000/Pk
Call For Price
COMPARE
0.5 Clear Colored PCR Tubes, Clear, 0.5 mL; 1000/Pk
Call For Price
Showing 1- 7 of 7