This product is now discontinued, View Related Products
PCRmax ®PCR Strip Tubes
Get quicker cycles times and rapid heat transfer with ultrathin walls
  • Constructed of pure high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
Suitable for use in common thermal cyclers, these strip tubes are available in PCR as well as qPCR options. Both options come in a strip of 8 connected tubes. The strips of PCR tubes are available in five color options, with corresponding domed cap strips sold separately. The strips of qPCR strips are available in one color and have corresponding flat cap strips sold separately.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Color Description
Availability Pricing
COMPARE
0.2 Clear PCR Strip Tubes, PP, 0.2 mL, Clear; 125/Cs
Call For Price
COMPARE
0.2 Clear PCR Strip Tubes, PP, 0.2 mL, Red; 125/Cs
Call For Price
COMPARE
0.2 Clear PCR Strip Tubes, PP, 0.2 mL, Yellow; 125/Cs
Call For Price
COMPARE
0.2 Clear PCR Strip Tubes, PP, 0.2 mL, Green; 125/Cs
Call For Price
COMPARE
0.2 Clear PCR Strip Tubes, PP, 0.2 mL, Blue; 125/Cs
Call For Price
COMPARE
0.2 White PCR/qPCR Strip Tubes, 0.2 mL, White; 125/Cs
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
Showing 1- 8 of 8