PCRmax® Polypropylene PCR Microplates
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results
  • Manufactured with 99.9% pure virgin polypropylene
  • RNAse/DNAse-free, nonpyrogenic, nonsterile
  • Compatible with sealing film
PCRmax® microplates are designed to fit real-time PCR thermal cyclers and sequencers. The construction of pure, clear polypropylene allows for full transparency. Microplates are compatible with PCRmax Alpha Cycler instruments.
$210.00 - $725.00USD / Pkg of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Well Volume (µL) Description
Availability Pricing
COMPARE
96 200 96-well Polypropylene PCR Microplate, Half Skirt, Clear, Nonsterile; 50/Pk
$210.00USD / Pkg of 50
COMPARE
96 100 96-well Polypropylene Flat Top PCR Microplate, Low Profile, No Skirt, Clear, Nonsterile; 100/Pk
$447.00USD / Pkg of 100
COMPARE
96 100 96-well Polypropylene Flat Top PCR Microplate, No Skirt, Clear, Nonsterile; 100/Pk
$447.00USD / Pkg of 100
COMPARE
384 100 384-well Polypropylene PCR Microplate, Full Skirt, Clear, Nonsterile; 100/Pk
$725.00USD / Pkg of 100