PEAK Scientific Genius SQ 24 Nitrogen Generators
Hassle-free nitrogen source for your Single Quad LC-MS systems
  • Produces an analytical-grade nitrogen supply
  • Variable flow rate up to 34 L/min and pressure up to 116 psi
  • Compact size easily fits under most lab benches
  • Integrated internal compressors for easy plug and play
Genius SQ 24 nitrogen gas generator is a cutting-edge evolution combining advanced technology with refined and robust engineering. It provides a high purity nitrogen source for any Single Quad LC-MS system and other mission-critical laboratory applications where performance and reliability are paramount. Only standing 24 in (61 cm) tall, the compact design fits under most lab benches. With flow rates up to 24 L/min, the Genius SQ 24 meets and exceeds the maximum flow rate required by any Single Quad LC-MS on the market.
Genius SQ 24 delivers a reliable, high quality gas – designed from the ground up to deliver maximum performance for labs that cannot afford to compromise. By design, Genius SQ 24 is simple to set up and use. However, as an added benefit, installation and training services are offered where dedicated Peak field service engineers will visit you to assist with getting you up and running with no stress – order 86443-41 in addition to your generator.
$22,113.00 - $25,320.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Power (VAC) Description
Availability Pricing
PEAK Scientific Genius SQ 24 Nitrogen Generator, 24 L/min; 230 VAC EW-86443-03 Mfr # 3301661
COMPARE
24000 230 Nitrogen Generator, 24 L/min; 120 VAC
$25,320.00USD / Each
PEAK Scientific Genius SQ 24 Nitrogen Generator, 24 L/min; 120 VAC EW-86443-04 Mfr # 3301660
COMPARE
24000 120 Nitrogen Generator, 24 L/min; 230 VAC
$22,113.00USD / Each
PEAK Scientific Genius SQ 24 Nitrogen Generator, 24 L/min; 100 VAC EW-86443-05 Mfr # 3301650
COMPARE
24000 100 Nitrogen Generator, 24 L/min; 100 VAC
$22,113.00USD / Each