PendoTECH® Pressure Monitoring and Transmitting Systems
Single-use solution to monitor process pressure
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
  • Data output capability to a PC or control system
  • User-configurable Min/Max alarm set points
The PressureMAT combines a pressure monitor, data logger, alarm, and transmitter in one simple control box. Designed for use with single-use, in-line pressure sensors. Each channel takes pressure readings once per second. Process pressure is displayed on the easy-to-read LCD. System is easily calibrated to zero through a single button. The PressureMAT may also transmit output via 4-20 mA signal or can provide RS-232 data transfer for use in common PC applications, such as the PressureMAT Data Acquisition Software or Excel®, through WinWedge or Hyper Terminal software.
14 PRODUCT OPTIONS
Compareitem specifications
Output Process Connection Description
Availability Pricing
PendoTech PressureMAT® Monitor/Transmitter, 2 Pressure Sensor Inputs, 2 Relay and 2 Analog Outputs or 4 Relay Outputs; 110/220 VAC EW-19406-02 Mfr # PMAT2
COMPARE
4 to 20 mA (2) and relays (2); or 4 relays 2 Pressure Inputs PMAT2 PressureMAT™ Monitor/Transmitter, 2 Pressure Sensor Inputs, 2 Relay and 2 Analog Outputs or 4 Relay Outputs; 110/220 VAC
$4,250.50USD / Each
PendoTech PressureMAT® Monitor/Transmitter, Displays in Bar, 2 Pressure Sensor Inputs, 2 Relay and 2 Analog Outputs; 110/220 VAC EW-19406-03 Mfr # PMAT2-BAR
COMPARE
4 to 20 mA (2) and relays (2); or 4 relays 2 Pressure Inputs PMAT2-BAR PressureMAT™ Monitor/Transmitter, Displays in Bar, 2 Pressure Sensor Inputs, 2 Relay and 2 Analog Outputs; 110/220 VAC
$3,724.50USD / Each
PendoTech PressureMAT® Monitor/Transmitter, 3 Pressure Sensor Inputs, 1 Relay and 3 Analog Outputs; 110/220 VAC
Always in Stock
EW-19406-04 Mfr # PMAT3
COMPARE
4 to 20 mA (3) and relay (1) 3 Pressure Inputs PMAT3 PressureMAT™ Monitor/Transmitter, 3 Pressure Sensor Inputs, 1 Relay and 3 Analog Outputs; 110/220 VAC
$4,139.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter, Displays in Bar, 3 Pressure Sensor Inputs, 1 Relay and 3 Analog Outputs; 110/220 VAC EW-19406-05 Mfr # PMAT3-BAR
COMPARE
4 to 20 mA (3) and relay (1) 3 Pressure Inputs PMAT3-BAR PressureMAT™ Monitor/Transmitter, Displays in Bar, 3 Pressure Sensor Inputs, 1 Relay and 3 Analog Outputs; 110/220 VAC
$4,254.50USD / Each
PendoTech PressureMAT® Monitor/Transmitter, 4 Pressure Sensor Inputs, 4 Analog Outputs; 110/220 VAC EW-19406-06 Mfr # PMAT4A
COMPARE
4 to 20 mA - 4 outputs 4 Pressure Inputs PMAT4A PressureMAT™ Monitor/Transmitter, 4 Pressure Sensor Inputs, 4 Analog Outputs; 110/220 VAC
$4,236.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter, Displays in Bar, 4 Pressure Sensor Inputs, 4 Analog Outputs; 110/220 VAC EW-19406-07 Mfr # PMAT4A-BAR
COMPARE
4 to 20 mA - 4 outputs 4 Pressure Inputs PMAT4A-BAR PressureMAT™ Monitor/Transmitter, Displays in Bar, 4 Pressure Sensor Inputs, 4 Analog Outputs; 110/220 VAC
$4,431.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter, 4 Pressure Sensor Inputs, 4 Relay Outputs; 110/220 VAC EW-19406-08 Mfr # PMAT4R
COMPARE
4 relays 4 Pressure Inputs PMAT4R PressureMAT™ Monitor/Transmitter, 4 Pressure Sensor Inputs, 4 Relay Outputs; 110/220 VAC
$5,392.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter, Displays in Bar, 4 Pressure Sensor Inputs, 4 Relay Outputs; 110/220 VAC EW-19406-09 Mfr # PMAT4R-BAR
COMPARE
4 relays 4 Pressure Inputs PMAT4R-BAR PressureMAT™ Monitor/Transmitter, Displays in Bar, 4 Pressure Sensor Inputs, 4 Relay Outputs; 110/220 VAC
$4,413.50USD / Each
PendoTech PressureMAT® Monitor/Transmitter, 3 Pressure Sensor Inputs, 1 Flow Sensor Input, and 4 Analog Outputs; 110/220 VAC EW-19406-10 Mfr # PMAT3P
COMPARE
4 to 20 mA - 4 outputs 3 Pressure, 1 Flow Inputs PMAT3P PressureMAT™ Monitor/Transmitter, 3 Pressure Sensor Inputs, 1 Flow Sensor Input, and 4 Analog Outputs; 110/220 VAC
$4,362.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter, Displays in Bar, 3 Pressure Sensor Inputs, 1 Flow Sensor Input, and 4 Analog Outputs; 110/220 VAC EW-19406-11 Mfr # PMAT3P-BAR
COMPARE
4 to 20 mA - 4 outputs 3 Pressure, 1 Flow Input PMAT3P-BAR PressureMAT™ Monitor/Transmitter, Displays in Bar, 3 Pressure Sensor Inputs, 1 Flow Sensor Input, and 4 Analog Outputs; 110/220 VAC
$4,457.50USD / Each
PendoTech PressureMAT® Monitor/Transmitter, Displays in Bar, 1 Pressure Sensor Input, 1 Relay and 1 Analog Output; 110/220 VAC EW-19406-40 Mfr # PMAT-S
COMPARE
4 to 20 mA (1) and relay (1) 1 Pressure Input PMAT-S PressureMAT™ Monitor/Transmitter, Displays in Bar, 1 Pressure Sensor Input, 1 Relay and 1 Analog Output; 110/220 VAC
$2,366.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter, 2 Pressure Sensor Inputs, 2 Flow Sensor Inputs, and 4 Analog Outputs; 110/220 VAC EW-19406-41 Mfr # PMAT-2P
COMPARE
4 to 20 mA - 4 outputs 2 Pressure, 1 Flow Input PMAT-2P PressureMAT™ Monitor/Transmitter, 2 Pressure Sensor Inputs, 2 Flow Sensor Inputs, and 4 Analog Outputs; 110/220 VAC
$4,278.00USD / Each
PendoTech PressureMAT® Monitor/Transmitter Bundle, Three Polysulfone Luer Sensors; 110/220 VAC
Cole-Parmer--
EW-19407-00 Mfr # MAKE-UP
COMPARE
4 to 20 mA (3) and relay (1) 3 Pressure Inputs PMAT3 PressureMAT™ Monitor/Transmitter Bundle, Three Polysulfone Luer Sensors; 110/220 VAC
$4,555.50USD / Each
PendoTech PressureMAT® Monitor/Transmitter Bundle, Three Polysulfone 1/4" Barb Sensors; 110/220 VAC
Cole-Parmer--
EW-19407-02 Mfr # MAKE-UP
COMPARE
4 to 20 mA (3) and relay (1) 3 Pressure Inputs PMAT3 PressureMAT™ Monitor/Transmitter Bundle, Three Polysulfone 1/4" Barb Sensors; 110/220 VAC
$4,598.50USD / Each
Showing 1- 14 of 14