PendoTECH® Single-Use Temperature Sensors
Eliminates cross-contamination and extensive cleaning protocols
  • Can be sterilized by gamma irradiation
  • High sensitivity to temperature change
  • Better than ±0.2°C accuracy in specified range of 0 to 70°C
  • Certificate of quality included
The PendoTECH single-use temperature sensors allow for highly accurate measuring of liquid temperature within a tube in a range of 0 to 70°C. Their unique design allows part of the product that comes in contact with the fluid to be disposable. The sensors eliminate cross-contamination and extensive cleaning protocols. Although designed for single-use applications, the sensors are robust enough to be repeatedly cleaned and reused.
The sensors are available with either a hose barb or luer fitting for easy connection to process tubing. A disposable temperature dip probe is also available to measure temperature within a vessel or water bath. The hose barb models are constructed of polysulfone while the luer sensor is constructed of polycarbonate. The temperature sensing element in both sensors is a thermistor embedded in a stainless steel tip. No calibration is required because the temperature versus resistance for the thermistor element is well-defined within the specified accuracy range.
The hose barb fitting sensor imparts no obstruction on the fluid path that can cause a pressure drop and there is no dead-leg at the point where the temperature is measured. The luer fitting sensor is ideal for lab scale development applications and can connect to a variety of fittings that can securely adapt to tubing or other devices.
These sensors can be connected to a wide range of monitors and transmitters that can read the resistance measurement from the thermistor and convert it to a temperature reading. A reusable cable with a ¼” headphone plug (19406-15) can connect the sensor directly to a suitable monitor such as the Digi-Sense® handheld thermometer (91428-04) or a temperature sensor transmitter (19406-45). The temperature sensor transmitter with a linear 4-20 mA output signal allows a temperature reading to be displayed in another location such as process controller. To transmit temperature readings to a data logger, three benchtop transmitter options (19406-47, -57 and -58) are also available.
The sensors all have a three-year shelf life and all product contact materials pass the USP Class VI testing with the exception of the polycarbonate connector for luer sensor and the temperature sensor dip probe.
$47.00 - $92.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° C) Description
Availability Pricing
PendoTech TEMPC-N-999 Single-Use Temperature Sensor, Polycarbonate, Luer Fitting EW-19406-12 Mfr # TEMPC-N-999
COMPARE
Luer Fittings 70 Single-Use Temperature Sensor, Polycarbonate, Luer Fitting
$84.00USD / Each
PendoTech TEMPS-N-050 Single-Use Temperature Sensor, Non-Sterile, Polysulfone; 1/2" Hose Barb EW-19406-13 Mfr # TEMPS-N-050
COMPARE
1/2" Hose Barb 70 Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/2" Hose Barb
$91.00USD / Each
PendoTech TEMPS-N-038 Single-Use Temperature Sensor, Non-Sterile, Polysulfone; 3/8" Hose Barb EW-19406-16 Mfr # TEMPS-N-038
COMPARE
3/8" Hose Barb 70 Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 3/8" Hose Barb
$85.00USD / Each
PendoTech TEMPS-N-025 Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/4" Hose Barb EW-19406-17 Mfr # TEMPS-N-025
COMPARE
1/4" Hose Barb 70 Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/4" Hose Barb
$85.00USD / Each
PendoTech TEMPS-N-075 Single-Use Temperature Sensor, Non-Sterile, Polysulfone; 3/4” Hose Barb EW-19406-42 Mfr # TEMPS-N-075
COMPARE
3/4" Hose Barb 70 Single-Use Temperature Sensor, Non-Sterile, Polysulfone 3/4” Hose Barb
$84.00USD / Each
PendoTech TEMPS-N-012 Single-Use Temperature Sensor, Non-Sterile, Polysulfone; 1/8” Hose Barb EW-19406-43 Mfr # TEMPS-N-012
COMPARE
1/8" Hose Barb 70 Single-Use Temperature Sensor, Non-Sterile, Polysulfone 1/8” Hose Barb
$92.00USD / Each
PendoTech TEMP-S-DP Single-Use Temperature Sensor Dip Probe Tip, Sterile; 3 ft EW-19406-44 Mfr # TEMP-S-DP
COMPARE
Molex 70 Single-Use Temperature Sensor Dip Probe Tip, Sterile, Molex Connector to Monitor Cable
$47.00USD / Each
Showing 1- 7 of 7