Penicillin-G Potassium Salt

  • Assay - ≥85%
  • pH of a 0.6% Solution - 5.5 to 7.5
  • Loss on Drying at 105°C - ≤1.0%
$130.00USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP914100 Penicillin-G Potassium Salt (100 million units) EW-88068-53 Mfr # BP914100
COMPARE
113-98-4
$130.00USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat