PFA Sanitary Clamp Tees
Ideal for extremely corrosive materials—PFA will not rouge or pit
  • Inert, nonstick surfaces—perfect for fragile and metal-sensitive proteins
  • Employ regular steam-in-place cycles without worry
  • Fully compatible with other Tri-Clamp® connections
  • Protective overmolding reinforces clamp connection sites
  • Full static vacuum capability at 300°F (149°C)
Sanitary applications that use highly corrosive chemicals require fittings that can endure the punishment. These food and pharmaceutical quality PFA fittings are easily cleaned so you will minimize downtime, cross contamination, and yield loss. Autoclavable.
$97.00 - $187.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in)
Availability Pricing
COMPARE
PFA 1/4
$97.00USD / Each
COMPARE
PFA 1/2
$106.00USD / Each
Venture Tech TE12 Sanitary Clamp Fittings, 3/4" Tri-Clamp Tee, PFA EW-31530-11 Mfr # TE12
COMPARE
PFA 3/4
$125.00USD / Each
Venture Tech TE16 Sanitary Clamp Fittings, 1" Tri-Clamp Tee, PFA EW-31530-12 Mfr # TE16
COMPARE
PFA 1
$150.00USD / Each
Venture Tech TE24 Tee connector, PFA, 1-1/2" sanitary clamp EW-31530-13 Mfr # TE24
COMPARE
PFA 1-1/2
$187.00USD / Each