PFA Straight Threaded Couplings, NPT (F)
Create a secure pipe threaded connection
  • Constructed of chemically inert PFA
These straight threaded coupling fittings are made of chemically inert PFA. Fittings can be sterilized via autoclave or ethylene oxide (ETO).
$34.00 - $107.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Thread size NPT(F) (port 1) (in) Thread size NPT(F) (port 2) (in)
Availability Pricing
COMPARE
PFA 1/8 1/8
$52.50USD / Each
COMPARE
PFA 1/4 1/4
$51.00USD / Each
COMPARE
PFA 3/8 3/8
$69.00USD / Each
COMPARE
PFA 1/2 1/2
$74.50USD / Each
COMPARE
PFA 3/4 3/4
$107.00USD / Each
Parker Hannifin PC-2-P Parker Straight Threaded Coupling, PFA, 1/8" NPT(F)
Buy More Save More
EW-31320-59 Mfr # PC-2-P
COMPARE
PFA 1/8 1/8
$34.00USD / Each
Parker Hannifin Parker Straight Threaded Coupling, PFA, 1/4" NPT(F)
Buy More Save More
EW-31320-60 Mfr # PC-4-P
COMPARE
PFA 1/4 1/4
$37.00USD / Each
Parker Hannifin Parker Straight Threaded Coupling, PFA, 3/8" NPT(F)
Buy More Save More
EW-31320-61 Mfr # PC-6-P
COMPARE
PFA 3/8 3/8
$51.00USD / Each
Parker Hannifin PC-8-P Parker Straight Threaded Coupling, PFA, 1/2" NPT(F)
Buy More Save More
EW-31320-62 Mfr # PC-8-P
COMPARE
PFA 1/2 1/2
$56.50USD / Each
Parker Hannifin PC-12-P Parker Straight Threaded Coupling, PFA, 3/4" NPT(F)
Buy More Save More
EW-31320-63 Mfr # PC-12-P
COMPARE
PFA 3/4 3/4
$73.00USD / Each
Showing 1- 10 of 10