PharMed® BPT Biocompatible Peristaltic Pump Tubing
Biocompatible inner bore is perfect for sensitive bioprocess fluids
  • Durable tubing provides long pump life, outlasting silicone tubing
  • Withstands repeated clean-in-place (CIP) and steam-in-place (SIP) cleaning and sterilization
  • Excellent gas barrier with very low gas permeability
  • Opaque tubing—perfect for light sensitive fluids
  • Good general chemical resistance
  • USP Class VI and USP <87> compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
A combination of biocompatible fluid surface and exceptional flex life makes PharMed BPT tubing ideal for transferring sensitive bioprocessing fluids. Formulated for use in the pharmaceutical and biotechnology industries, it is ideal for use in cell harvesting, media processing, vaccine manufacturing, aseptic filling, and diagnostic testing applications.
The PharMed BPT tubing withstands repeated CIP and SIP, simplifying cleaning and sterilization. Tubing can be used with most commercial cleaners and sanitizers; autoclave up to five cycles or use gamma irradiation without affecting overall service life.
This specially formulated tubing features exceptional flex life characteristics and excellent wear properties to outlast silicone in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability characteristics provide protection for sensitive fluids from gas ingress. Opaque tubing protects light-sensitive fluids from visible and UV lights.
$90.30 - $557.00USD / Pkg of 1
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/50" ID x 9/64" OD; 25 Ft EW-50123-49 Mfr # AY242605
COMPARE
1/50 9/64 1/16
$90.30USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/32" ID x 5/32" OD; 25 Ft EW-50123-50 Mfr # AY242606
COMPARE
1/32 5/32 1/16
$107.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/16" ID x 1/8" OD; 25 Ft EW-50123-51 Mfr # AY242002
COMPARE
1/16 1/8 1/32
$119.25USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/16" ID x 3/16" OD; 25 Ft EW-50123-52 Mfr # AY242003
COMPARE
1/16 3/16 1/16
$133.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/32" ID x 7/32" OD; 25 Ft EW-50123-53 Mfr # AY242005
COMPARE
3/32 7/32 1/16
$172.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/8" ID x 3/16" OD; 25 Ft EW-50123-54 Mfr # AY242006
COMPARE
1/8 3/16 1/32
$130.75USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/8" ID x 1/4" OD; 25 Ft EW-50123-55 Mfr # AY242007
COMPARE
1/8 1/4 1/16
$138.25USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/16" ID x 5/16" OD; 25 Ft EW-50123-56 Mfr # AY242012
COMPARE
3/16 5/16 1/16
$141.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/16" ID x 3/8" OD; 25 Ft EW-50123-57 Mfr # AY242013
COMPARE
3/16 3/8 3/32
$144.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/4" ID x 3/8" OD; 25 Ft EW-50123-58 Mfr # AY242017
COMPARE
1/4 3/8 1/16
$167.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/4" ID x 1/2" OD; 25 Ft EW-50123-59 Mfr # AY242019
COMPARE
1/4 1/2 1/8
$293.50USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 5/16" ID x 7/16" OD; 25 Ft EW-50123-60 Mfr # AY242022
COMPARE
5/16 7/16 1/16
$175.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/8" ID x 1/2" OD; 25 Ft EW-50123-61 Mfr # AY242027
COMPARE
3/8 1/2 1/16
$214.00USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/8" ID x 5/8" OD; 25 Ft EW-50123-62 Mfr # AY242029
COMPARE
3/8 5/8 1/8
$276.25USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 1/2" ID x 3/4" OD; 25 Ft EW-50123-63 Mfr # AY242038
COMPARE
1/2 3/4 1/8
$337.75USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 5/8" ID x 7/8" OD; 25 Ft EW-50123-64 Mfr # AY242046
COMPARE
5/8 7/8 1/8
$399.25USD / Pkg of 1
PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/4" ID x 1" OD; 25 Ft EW-50123-65 Mfr # AY242053
COMPARE
3/4 1 1/8
$557.00USD / Pkg of 1
Showing 1- 17 of 17