This product is now discontinued, View Related Products
Phase II Digital Coating Thickness Gauges
Provides non-destructive precision accurate testing performance
  • Calculates thickness measurement using eddy current and magnetic induction
  • Compact, durable design for industrial environments
  • Integrated or eternal probe configurations
These precision coating thickness gauges allow you to easily detect the thickness of non-magnetic coating on a magnetic substrate (ferrous) or an insulating coating on a non-magnetic conductive substrate (non-ferrous) utilizing either an integrated probe (model PTG-3700) or external probe (model PGT-3750). Thickness coating gauges are ideal for applications that include automotive paint measurement, manufacturing, general engineering, commercial inspection, and more.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Measuring Method Probe Range
Availability Pricing
Phase-II PTG-3700 Coating Thickness Gauge with internal probe EW-54102-15 Mfr # PTG-3700
COMPARE
0 to 50 mil (0 to 1250 µm)
Call For Price
Phase-II PTG-3750 Coating Thickness Gauge with external probe EW-54102-16 Mfr # PTG-3750
COMPARE
Eddy current, Magnetic induction 0 to 50 mil (0 to 1250 µm)