This product is now discontinued, View Related Products

Phenethyl alcohol

Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC130171000 Phenethyl alcohol 99+% (100g) EW-88132-17 Mfr # AC130171000
COMPARE
60-12-8
Call For Price
Acros Organics AC130172500 Phenethyl alcohol 99+% (250g) EW-88132-18 Mfr # AC130172500
COMPARE
60-12-8
Call For Price
Acros Organics AC130180010 Phenethyl alcohol 99% (1l) EW-88132-19 Mfr # AC130180010
COMPARE
60-12-8
Call For Price
Acros Organics AC130180025 Phenethyl alcohol 99% (2.5l) EW-88132-20 Mfr # AC130180025
COMPARE
60-12-8
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat