Phosphate Buffered Saline, 10X Solution

  • pH of a 10X Solution at 25°C - 6.7 to 6.9
  • pH of a 1X Solution at 25°C - 7.3 to 7.5
  • Conductivity of a 1X Solution - 14,000 to 17,800μmhos
  • Chloride Concentration of a 1X Solution - 0.140 ±0.004 moles/L
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected
$56.50 - $698.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher BioReagents BP3991 PBS, 10X Solution (1 liter) CAS 7647-14-5 EW-88067-22 Mfr # BP3991
COMPARE
7647145 (Sodium Chloride) 7447407 (Potassium Chloride) 7558794 (Sodium Phosphate Dibasic) 7778770 (Potassium Phosphate Monobasic) 1000 PBS, 10X Solution (1 liter) CAS 7647-14-5
$99.00USD / Each
Fisher BioReagents BP39920 PBS, 10X Solution (20 liter) CAS 7647-14-5 EW-88067-23 Mfr # BP39920
COMPARE
7647145 (Sodium Chloride) 7447407 (Potassium Chloride) 7558794 (Sodium Phosphate Dibasic) 7778770 (Potassium Phosphate Monobasic) 20000 PBS, 10X Solution (20 liter) CAS 7647-14-5
$698.00USD / Each
Fisher BioReagents BP3994 PBS, 10X Solution (4 liter) CAS 7647-14-5 EW-88067-24 Mfr # BP3994
COMPARE
7647145 (Sodium Chloride) 7447407 (Potassium Chloride) 7558794 (Sodium Phosphate Dibasic) 7778770 (Potassium Phosphate Monobasic) 4000 PBS, 10X Solution (4 liter) CAS 7647-14-5
$214.00USD / Each
Fisher BioReagents BP399500 PBS, 10X Solution (500 mL) CAS 7647-14-5 (PBS) EW-88067-25 Mfr # BP399500
COMPARE
7647145 (Sodium Chloride) 7447407 (Potassium Chloride) 7558794 (Sodium Phosphate Dibasic) 7778770 (Potassium Phosphate Monobasic) 500 PBS, 10X Solution (500 mL) CAS 7647-14-5 (PBS)
$56.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat