This product is now discontinued, View Related Products

Fisher BioReagents™ Phosphate Buffered Saline, 1X Powder Concentrate

Reagents and solutions to meet your lab and research needs
  • Solubility (1% in water) - To pass test
  • pH of a 1X Solution at 25°C - 7.3 to 7.5
  • Conductivity of a 1X Solution - 14,000 to 17,800μmhos
  • Chloride Concentration of a 1X Solution - 0.140±0.004 moles/L
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected
Quality and consistency your lab can count on.
Call For Price

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
Fisher BioReagents BP66150 PBS, 1X Powder Concentrate (White Granular Powder) (50 liter) EW-88068-27 Mfr # BP66150
COMPARE
50000 PBS, 1X Powder Concentrate (White Granular Powder) (50 liter)
Call For Price
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat