This product is now discontinued, View Related Products

Phosphorus Oxychloride

  • Heavy Metals (as Pb) - ≤0.002%
  • Iron - ≤0.001%
  • Sulfate - ≤0.01%
  • Boiling Range - 105°-109°C
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical P106I500 Phosphorus Oxychloride (Certified) (500mL) EW-88078-27 Mfr # P106I500
COMPARE
10025-87-3
Call For Price
Acros Organics AC191290010 Phosphorus oxychloride 99% (1kg) CAS 10025-87-3 EW-88204-35 Mfr # AC191290010
COMPARE
10025-87-3
Call For Price
Acros Organics AC191290050 Phosphorus oxychloride 99% (5g) EW-88204-36 Mfr # AC191290050
COMPARE
10025-87-3
Call For Price
Acros Organics AC191292500 Phosphorus oxychloride 99% (250g) EW-88204-37 Mfr # AC191292500
COMPARE
10025-87-3
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat