Plastic Hand-Operated Heavy-Duty Acid-Transfer Hand Pumps
Ideal for most acid/base or ionic salt applications
  • UL-listed for sulfuric acid transfer at 50% concentration
  • Withstand temperatures up to 120°>F (49°>C)
  • Self-priming, will not siphon
These plastic hand-operated drum pumps are ideal for heavy-duty applications that require acid-transfer. All models are constructed of solvent-welded PVC body and shaft with Viton® valves and seals and feature 6 strokes/gallon (2 strokes/liter).
$182.40 - $308.70USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Container Adapter Flow Rate (Strokes/Gal)
Availability Pricing
COMPARE
Glass Carboy 6
$308.70USD / Each
COMPARE
None 6
$182.40USD / Each
COMPARE
2" IPS 6
$194.60USD / Each
COMPARE
Buttress Dell Drum (M) 6
$192.20USD / Each
COMPARE
83 mm Buttress (F) 6
$267.40USD / Each