Plastic Needle Valves
PVDF needle valves are pressure rated up to 200 psi (13.8 bar) with a max temperature rating of 302 deg F (150 deg C). Seats are made of PTFE and O-rings made of Viton®. Valves feature standard pipe thread connections and are autoclavable.
$184.00 - $197.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
GF Signet 155.522.554 PVDF Needle Valve, 1/4" NPT(F), with PTFE EW-01369-20 Mfr # 155.522.554
COMPARE
1/4" NPT(F) 1/4" NPT(F) PVDF
$197.00USD / Each
GF Signet 155.522.554 PVDF Needle Valve, 3/8" NPT(F), with PTFE and Viton EW-01369-21 Mfr # 155.522.555
COMPARE
3/8" NPT(F) 3/8" NPT(F) PVDF
$184.00USD / Each
GF Signet 155.522.556 PVDF Needle Valve, 1/2" NPT(F), with PTFE EW-01369-22 Mfr # 155.522.556
COMPARE
1/2" NPT(F) 1/2" NPT(F) PVDF
$190.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat