Pleated Carbon-Impregnated Cartridge
Non-cellulose media—resistant to bacterial attack and biofilm buildup
Pleated Carbon-Impregnated Cartridges provide high dirt-loading capacities and extended life. Non-cellulose carbon-impregnated media resist bacterial attack and biofilm buildup — use it in non-chlorinated, well water, and post-reverse osmosis applications.
Cartridge features powder carbon to reduce chlorine taste/odor and to reduce sediment particles down to 10 µm. Chlorine reduction‡‡ is approximately 1300 gallons at 1 GPM. Surface area is 4.5 sq ft. Maximum temperature is 150°F (66°C).
$17.10 - $32.75USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
Pentair 155367-43 Double open end pleated carbon-impregnated cartridge; 10 µm, 10"L EW-29830-30 Mfr # 155367-43
COMPARE
10 3 GPM 10
$17.10USD / Each
Pentair 155397-43 Double open end pleated carbon-impregnated cartridge; 10 µm, 20"L EW-29830-31 Mfr # 155397-43
COMPARE
20 5 GPM 10
$32.75USD / Each