Pleated Cartridges
Design offers higher flow rates and dirt-loading capacities than spun or wound depth cartridges, especially when removing lower micron-sized particles
  • Remove non-gelatinous particles that fall into a narrow size range, typically in post filtration or polishing applications
Pleated cartridges separate viscous sediments and flat particulates. Use polyester cartridges for water filtration and polypropylene cartridges with harsher chemicals. Plastisol end caps.
$24.00 - $56.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Filter Media Micron Rating
Availability Pricing
Big Blue 155120-43 Filter cartridge, "", 10", pleated polypropylene, 30 µm
Always in Stock
EW-01508-86 Mfr # 155120-43
COMPARE
10 Polypropylene 30
$40.50USD / Each
Big Blue Filter cartridge, 10", pleated cellulose, 20 µm EW-01512-74 Mfr # 155405-43(S1-BB)
COMPARE
10 Cellulose 20
$24.00USD / Each
Big Blue Filter cartridge, 20", pleated cellulose, 20 micron EW-01512-76 Mfr # 155305-43(S1-20BB)
COMPARE
20 Cellulose 20
$52.00USD / Each
Big Blue 255494-43 Filter cartridge, 20", pleated cellulose-polyester, 5 micron EW-01512-88 Mfr # 255494-43
COMPARE
20 Cellulose-polyester 5
$56.00USD / Each
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat