Plug Fittings, Male NPT Threaded, Straight
Deigned for permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).
$24.50 - $96.50USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Thread size NPT(M) (port 1) (in)
Availability Pricing
COMPARE
PFA NPT(M) 1/8
$27.25USD / Each
COMPARE
PFA NPT(M) 1/4
$29.75USD / Each
COMPARE
PFA NPT(M) 3/8
$24.50USD / Each
COMPARE
PFA NPT(M) 1/2
$43.50USD / Each
COMPARE
PFA NPT(M) 3/4
$47.75USD / Each
COMPARE
PTFE NPT(M) 1/8
$38.00USD / Each
COMPARE
PTFE NPT(M) 1/4
$47.75USD / Each
COMPARE
PTFE NPT(M) 3/8
$48.25USD / Each
COMPARE
PTFE NPT(M) 1/2
$56.00USD / Each
COMPARE
PTFE NPT(M) 3/4
$70.00USD / Each
COMPARE
PTFE NPT(M) 1
$96.50USD / Each
Showing 1- 11 of 11