Fisher BioReagents™ Polyethylene Glycol 8000
Suitable as a medium for fusion of mammalian cells
  • Reagents and solutions to meet your lab and research needs
The polyethylene glycol 8000 is suitable as a medium for the fusion of mammalian cells. It may also be used to precipitate bacteriophage from lysed cell supernatants.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$157.00 - $177.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Packaging Weight (g) Description
Availability Pricing
Fisher BioReagents Polyethylene Glycol 8000, 1 kg EW-88058-53 Mfr # BP2331
COMPARE
Poly Bottle 1000 Polyethylene Glycol 8000, 1 kg
$177.50USD / Each
Fisher BioReagents Polyethylene Glycol 8000, 100 g EW-88058-54 Mfr # BP233100
COMPARE
Glass Bottle 100 Polyethylene Glycol 8000, 100 g
$157.00USD / Each