Polyfoil Square Wrap
Polyester laminated foil is resistant to punctures, tears and corrosion. Ideal for use with organic solvents and caustic solutions. Easy to apply and remove, folds easily, and resists kinking. Autoclavable. 100 per pack.
$85.15USD / of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Material Thickness (mil)
Availability Pricing
Polyfoil Polyester Lined Aluminum Foil, 8 x 8" Squares; 100/Box
Buy More Save More
EW-06275-06 Mfr # F13200-0088
COMPARE
Polyester-lined Aluminum
$85.15USD / of 1