Polypropylene Adapter Heads 18362
Attach bags onto any open system drain or sump-return to collect solids
  • Solids are contained within the bag for easy disposal
An improvement over tie-on designs—the head and support basket make the full filter bag surface area available for use. Filter bags include a galvanized steel ring sewn into the top that
securely holds the bag to the head. Polypropylene bags are glazed to reduce fiber migration and have welded seams for high strength. All bag designs are suitable to 100°F (38°C).
$4.45USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Filter Media Micron Rating
Availability Pricing
COMPARE
8 Polypropylene 10
$4.45USD / Each