Polypropylene and PVC Pressure Regulators
Easily control your outlet pressure
  • Choose between PVC or polypropylene (PP) construction with Viton® diaphragm
Valves feature a diaphragm made of Viton®. Use to control your outlet pressure between 5 and 50 psig; up to 150 psig inlet. Ideal for your liquid applications.
$698.00 - $850.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
COMPARE
1/2" NPT(F) 1/2" NPT(F) PVC
$698.00USD / Each
COMPARE
1/2" NPT(F) 1/2" NPT(F) Polypropylene
$850.00USD / Each