Polypropylene Bag Filter Housings
  • Polypropylene construction provides corrosion resistance, strength, and durability
  • Housings are lightweight and portable
  • Maximum temperature is 100°F (38°C)
Polypropylene is ideal for water, acids, plating solutions, corrosive chemicals, food products, and more. Each housing includes a Buna-N O-ring, gauge, wrench, ¼" vent port with plug, and 3/8" drain valve.
$222.00 - $252.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Flow Rate Port Size
Availability Pricing
COMPARE
10 20 GPM 1" NPT(F)
$252.00USD / Each
COMPARE
10 40 GPM 1" NPT(F)
$250.00USD / Each
COMPARE
10 20 GPM 1.5" NPT(F)
$222.00USD / Each
COMPARE
10 40 GPM 1.5" NPT(F)
$249.00USD / Each