Polypropylene Hose Barb Inserts
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
$2.95 - $15.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) PLCD22004 Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/4" Flow Size, Acetal
Buy More Save More
EW-06360-80 Mfr # PLCD22004
COMPARE
1/4 Acetal Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/4" Flow Size, Acetal
$8.40USD / Each
CPC (Colder) PLC22004 Quick-Disconnect Hose Barb Insert; Straight-through, 1/4" Tube ID, Acetal
Buy More Save More
EW-06360-82 Mfr # PLC22004
COMPARE
1/4 Acetal Quick-Disconnect Hose Barb Insert; Straight-through, 1/4" Tube ID, Acetal
$2.95USD / Each
CPC (Colder) PLCD22005 Quick-Disconnect Hose Barb Insert; Valved, 5/16" Tube ID, 1/4" Flow Size
Buy More Save More
EW-06360-83 Mfr # PLCD22005
COMPARE
5/16 Acetal Quick-Disconnect Hose Barb Insert; Valved, 5/16" Tube ID, 1/4" Flow Size
$8.50USD / Each
CPC (Colder) PLC22005 Quick-Disconnect Hose Barb Insert; Straight-through, 5/16" Tube ID, Acetal
Buy More Save More
EW-06360-84 Mfr # PLC22005
COMPARE
5/16 Acetal Quick-Disconnect Hose Barb Insert; Straight-through, 5/16" Tube ID, Acetal
$3.35USD / Each
CPC (Colder) PLCD22006 Quick-Disconnect Hose Barb Insert; Valved, 3/8" Tube ID, 1/4" Flow Size, Acetal
Buy More Save More
EW-06360-85 Mfr # PLCD22006
COMPARE
3/8 Acetal Quick-Disconnect Hose Barb Insert; Valved, 3/8" Tube ID, 1/4" Flow Size, Acetal
$8.50USD / Each
CPC (Colder) PLC22006 Quick-Disconnect Hose Barb Insert; Straight-through, 3/8" Tube ID, Acetal
Buy More Save More
EW-06360-87 Mfr # PLC22006
COMPARE
3/8 Acetal Quick-Disconnect Hose Barb Insert; Straight-through, 3/8" Tube ID, Acetal
$3.20USD / Each
CPC (Colder) PLCD2200412 Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/4" Flow Size, PP
Buy More Save More
EW-06364-80 Mfr # PLCD2200412
COMPARE
1/4 Polypropylene Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/4" Flow Size, PP
$12.50USD / Each
CPC (Colder) PLC2200412 Quick-Disconnect Hose Barb Insert; Straight-through, 1/4" Tube ID, PP
Buy More Save More
EW-06364-82 Mfr # PLC2200412
COMPARE
1/4 Polypropylene Quick-Disconnect Hose Barb Insert; Straight-through, 1/4" Tube ID, PP
$4.90USD / Each
CPC (Colder) PLCD2200612 Quick-Disconnect Hose Barb Insert; Valved, 3/8" Tube ID, 1/4" Flow Size, PP
Buy More Save More
EW-06364-85 Mfr # PLCD2200612
COMPARE
3/8 Polypropylene Quick-Disconnect Hose Barb Insert; Valved, 3/8" Tube ID, 1/4" Flow Size, PP
$15.00USD / Each
CPC (Colder) PLC2200612 Quick-Disconnect Hose Barb Insert; Straight-through, 3/8" Tube ID, PP
Buy More Save More
EW-06364-87 Mfr # PLC2200612
COMPARE
3/8 Polypropylene Quick-Disconnect Hose Barb Insert; Straight-through, 3/8" Tube ID, PP
$4.60USD / Each
Showing 1- 10 of 10