Polypropylene (PP) Moderate-Flow Chemical-Resistance Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.
$180.80 - $846.50USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Power (VAC)
Availability Pricing
COMPARE
2.3 4.5 115
$183.00USD / Each
COMPARE
2.3 4.5 220
$185.40USD / Each
COMPARE
3.1 6.8 115
$180.80USD / Each
COMPARE
3.1 6.8 220
$183.20USD / Each
COMPARE
5 11.1 115
$299.00USD / Each
COMPARE
5 11.1 220
$299.00USD / Each
COMPARE
8.2 14.1 115
$352.30USD / Each
COMPARE
8.2 14.1 220
$333.30USD / Each
COMPARE
10 17.7 115
$461.90USD / Each
COMPARE
10 17.7 220
$464.10USD / Each
COMPARE
13.7 21.3 115
$555.75USD / Each
COMPARE
13.7 21.3 220
$561.50USD / Each
COMPARE
18.4 26.9 115
$623.50USD / Each
COMPARE
18.4 26.9 220
$624.75USD / Each
COMPARE
25.6 31.8 115
$702.50USD / Each
COMPARE
25.6 31.8 220
$675.00USD / Each
COMPARE
35.6 39 115
$846.50USD / Each
COMPARE
35.6 39 220
$824.25USD / Each
Showing 1- 18 of 18