Polypropylene Sediment Cartridges
Polypropylene fiber medium is thermally bonded to a rigid core for added durability
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 175°F (79°C).
Note: For optimal performance, change cartridges at 25 psid.
$9.60 - $17.10USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
COMPARE
10 2 GPM at <2psid 1
$17.10USD / Each
COMPARE
20 5 GPM at <2 psid 1
$16.60USD / Each
COMPARE
10 2 GPM at <2 psid 5
$12.90USD / Each
COMPARE
20 5 GPM at <2 psid 5
$17.10USD / Each
COMPARE
10 2 GPM at 2 psid 10
$12.60USD / Each
COMPARE
20 5 GPM at <2 psid 10
$17.10USD / Each
COMPARE
10 2 GPM at <2 psid 25
$16.10USD / Each
COMPARE
20 5 GPM at <2 psid 25
$16.10USD / Each
COMPARE
10 2 GPM at <2 psid 50
$9.60USD / Each
COMPARE
20 5 GPM at <2 psid 50
$15.50USD / Each
Showing 1- 10 of 10