Polypropylene String-Wound Cartridges
String-wound pattern provides greater surface area for lower pressure drop and higher flow
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
$41.00 - $105.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Micron Rating Description
Availability Pricing
COMPARE
10 5 Fibrous polypropylene string-wound cartridge, 5 µm, 10" length
$44.50USD / Each
COMPARE
10 1 Fibrous polypropylene string-wound cartridge, 1 µm, 10" length
$42.50USD / Each
COMPARE
10 0.5 Fibrous polypropylene string-wound cartridge, 0.5 µm, 10" length
$48.50USD / Each
Pentair 355215-43 Fibrous polypropylene string-wound cartridge, 10 µm, 10" length EW-29802-35 Mfr # 355215-43
COMPARE
10 10 Fibrous polypropylene string-wound cartridge, 10 µm, 10" length
$41.00USD / Each
Pentair WP25BB97P Fibrous polypropylene string-wound cartridge, 25 µm, 10" length EW-29802-36 Mfr # WP25BB97P
COMPARE
10 25 Fibrous polypropylene string-wound cartridge, 25 µm, 10" length
$41.00USD / Each
Pentair WP1BB20P Fibrous polypropylene string-wound cartridge, 1 µm, 20" length EW-29802-37 Mfr # WP1BB20P
COMPARE
20 1 Fibrous polypropylene string-wound cartridge, 1 µm, 20" length
$105.00USD / Each
Pentair WP5BB20P Fibrous polypropylene string-wound cartridge, 5 µm, 20" length EW-29802-38 Mfr # WP5BB20P
COMPARE
20 5 Fibrous polypropylene string-wound cartridge, 5 µm, 20" length
$91.00USD / Each
Pentair WP25BB20P Fibrous polypropylene string-wound cartridge, 25 µm, 20" length EW-29802-39 Mfr # WP25BB20P
COMPARE
20 25 Fibrous polypropylene string-wound cartridge, 25 µm, 20" length
$89.50USD / Each
Pentair WPX5BB97P Fibrillated polypropylene string-wound cartridge, 5 µm, 10" length EW-29802-40 Mfr # WPX5BB97P
COMPARE
10 5 Fibrillated polypropylene string-wound cartridge, 5 µm, 10" length
$46.50USD / Each
Pentair WPX25BB97P Fibrillated polypropylene string-wound cartridge, 25 µm, 10" length EW-29802-42 Mfr # WPX25BB97P
COMPARE
10 25 Fibrillated polypropylene string-wound cartridge, 25 µm, 10" length
$46.00USD / Each
Pentair WPX50BB97P Fibrillated polypropylene string-wound cartridge, 50 µm, 10" length EW-29802-43 Mfr # WPX50BB97P
COMPARE
10 50 Fibrillated polypropylene string-wound cartridge, 50 µm, 10" length
$45.50USD / Each
Pentair WPX100BB97P Fibrillated polypropylene string-wound cartridge, 100 µm, 10" length EW-29802-44 Mfr # 355221-43
COMPARE
10 100 Fibrillated polypropylene string-wound cartridge, 100 µm, 10" length
$54.50USD / Each
Pentair WPX100BB20P Fibrillated polypropylene string-wound cartridge, 100 µm, 20" length EW-29802-45 Mfr # WPX100BB20P
COMPARE
20 100 Fibrillated polypropylene string-wound cartridge, 100 µm, 20" length
$81.50USD / Each
Showing 1- 13 of 13
Need Help with this product?
Call Us At1-847-549-7600
Live chat